Merck
  • Spastin tethers lipid droplets to peroxisomes and directs fatty acid trafficking through ESCRT-III.
Spastin tethers lipid droplets to peroxisomes and directs fatty acid trafficking through ESCRT-III.
The Journal of cell biology (2019-06-23)
Chi-Lun Chang, Aubrey V Weigel, Maria S Ioannou, H Amalia Pasolli, C Shan Xu, David R Peale, Gleb Shtengel, Melanie Freeman, Harald F Hess, Craig Blackstone, Jennifer Lippincott-Schwartz
ABSTRACT
Lipid droplets (LDs) are neutral lipid storage organelles that transfer lipids to various organelles including peroxisomes. Here, we show that the hereditary spastic paraplegia protein M1 Spastin, a membrane-bound AAA ATPase found on LDs, coordinates fatty acid (FA) trafficking from LDs to peroxisomes through two interrelated mechanisms. First, M1 Spastin forms a tethering complex with peroxisomal ABCD1 to promote LD-peroxisome contact formation. Second, M1 Spastin recruits the membrane-shaping ESCRT-III proteins IST1 and CHMP1B to LDs via its MIT domain to facilitate LD-to-peroxisome FA trafficking, possibly through IST1- and CHMP1B-dependent modifications in LD membrane morphology. Furthermore, LD-to-peroxisome FA trafficking mediated by M1 Spastin is required to relieve LDs of lipid peroxidation. M1 Spastin's dual roles in tethering LDs to peroxisomes and in recruiting ESCRT-III components to LD-peroxisome contact sites for FA trafficking may underlie the pathogenesis of diseases associated with defective FA metabolism in LDs and peroxisomes.
Sigma-Aldrich
Anti-Spastin Antibody, serum, from rabbit
Sigma-Aldrich
Anti-ACBD5 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution