Merck
  • High-Frequency Microdomain Ca2+ Transients and Waves during Early Myelin Internode Remodeling.
High-Frequency Microdomain Ca2+ Transients and Waves during Early Myelin Internode Remodeling.
Cell reports (2019-01-04)
Arne Battefeld, Marko A Popovic, Sharon I de Vries, Maarten H P Kole
ABSTRACT
Ensheathment of axons by myelin is a highly complex and multi-cellular process. Cytosolic calcium (Ca2+) changes in the myelin sheath have been implicated in myelin synthesis, but the source of this Ca2+ and the role of neuronal activity is not well understood. Using one-photon Ca2+ imaging, we investigated myelin sheath formation in the mouse somatosensory cortex and found a high rate of spontaneous microdomain Ca2+ transients and large-amplitude Ca2+ waves propagating along the internode. The frequency of Ca2+ transients and waves rapidly declines with maturation and reactivates during remyelination. Unexpectedly, myelin microdomain Ca2+ transients occur independent of neuronal action potential generation or network activity but are nearly completely abolished when the mitochondrial permeability transition pores are blocked. These findings are supported by the discovery of mitochondria organelles in non-compacted myelin. Together, the results suggest that myelin microdomain Ca2+ signals are cell-autonomously driven by high activity of mitochondria during myelin remodeling.
Sigma-Aldrich
SR-95531, ≥98% (HPLC), powder
Sigma-Aldrich
Anti-Proteolipid Protein Antibody, Chemicon®, from chicken
Sigma-Aldrich
Anti-Myelin Oligodendrocyte Glycoprotein (MOG) Antibody, clone 8-18C5, Chemicon®, from mouse