![Tryptophan European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/266/242/80aa41ff-217d-4f52-bf79-cbc2d23e4818/640/80aa41ff-217d-4f52-bf79-cbc2d23e4818.png)
Tryptophan
![Tryptophan European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/266/242/80aa41ff-217d-4f52-bf79-cbc2d23e4818/640/80aa41ff-217d-4f52-bf79-cbc2d23e4818.png)
Synonym(s):
L-Tryptophan, (S)-2-Amino-3-(3-indolyl)propionic acid, L-α-Amino-3-indolepropionic acid
Empirical Formula (Hill Notation):
C11H12N2O2
CAS Number:
Molecular Weight:
204.23
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing