Skip to Content
Merck
TAPS BioXtra, ≥99.5% (titration)

TAPS

TAPS BioXtra, ≥99.5% (titration)
Synonym(s):
N-[Tris(hydroxymethyl)methyl]-3-aminopropanesulfonic acid, [(2-Hydroxy-1,1-bis(hydroxymethyl)ethyl)amino]-1-propanesulfonic acid
Empirical Formula (Hill Notation):
C7H17NO6S
CAS Number:
Molecular Weight:
243.28
EC Number:

Products

Product Category
Brand
Feature
Formula Weight
Melting Point (°C)
pH Value
Physical Form
pK Value
Product Line
Purity
Quality Segment
Application
Showing 1-2 of 2
Compare
Product Number
Description
Pricing