
Rivastigmine Related Compound D

Synonym(s):
N-Ethyl-N-methylcarbamic acid 3-acetylphenyl ester, 3-Acetylphenyl ethyl(methyl)carbamate
Empirical Formula (Hill Notation):
C12H15NO3
CAS Number:
Molecular Weight:
221.25
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2