
(R)-(−)-1-Benzyloxy-2-propanol

Synonym(s):
(R)-1-O-Benzylpropylene glycol
Linear Formula:
C6H5CH2OCH2CH(OH)CH3
CAS Number:
Molecular Weight:
166.22
Products
Product Category
Brand
Formula Weight
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing