
Picroside I

Synonym(s):
(1aS,1bS,2S,5aR,6S,6aS)-1a,1b,2,5a,6,6a-Hexahydro-6-hydroxy-1a-(hydroxymethyl)oxireno[4,5]cyclopenta[1,2-c]pyran-2-yl-β-D-glucopyranoside 6-[(2E)-3-phenyl-2-propenoate], 6′-Cinnamoylcatalpol
Empirical Formula (Hill Notation):
C24H28O11
CAS Number:
Molecular Weight:
492.47
Products
Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Research Area
Quality Segment
Application
Optical Isomer
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing