
Phenazine ethosulfate

Synonym(s):
N-Ethyldibenzopyrazine ethyl sulfate salt
Linear Formula:
C16H18N2SO4
Products
Product Category
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Application
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing