
Petunidin 3-glucoside chloride

Synonym(s):
Petunidin 3-O-β-D-glucoside chloride, Petunidin 3-O-β-D-glucopyranoside, Petunidin 3-glucoside, Petunidin 3-glucoside chloride
Empirical Formula (Hill Notation):
C22H23ClO12
CAS Number:
Molecular Weight:
514.86
Products
Formula Weight
Manufacturer
Product Line
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing