
Pelargonidin 3-glucoside chloride

Synonym(s):
Callistephin chloride, 3-(Glucosyloxy)-4′,5,7-trihydroxyflavylium chloride, Pelargonidin 3-O-glucoside chloride
Empirical Formula (Hill Notation):
C21H21ClO10
CAS Number:
Molecular Weight:
468.84
Products
Formula Weight
Manufacturer
Product Line
Purity
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing