
NADP

Synonym(s):
β-Nicotinamide adenine dinucleotide phosphate disodium salt, β-NADH phosphate disodium salt, β-NADP-Na2, Triphosphopyridine nucleotide disodium salt
Empirical Formula (Hill Notation):
C21H26N7Na2O17P3
CAS Number:
Molecular Weight:
787.37
Products
Formula Weight
Manufacturer
Physical Form
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing