Skip to Content
Merck
N-Acetyl-L-phenylalanine ReagentPlus®, 99%

N-Acetyl-L-phenylalanine

N-Acetyl-L-phenylalanine ReagentPlus®, 99%
Linear Formula:
C6H5CH2CH(NHCOCH3)CO2H
CAS Number:
Molecular Weight:
207.23
EC Number:

Products

Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Amino Acid Type
Reaction Type
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing