
Loxapine Related Compound A

Synonym(s):
3-Chloro-11-(4-methylpiperazin-1-yl)dibenzo[b,f][1,4]oxazepine
Empirical Formula (Hill Notation):
C18H18ClN3O
CAS Number:
Molecular Weight:
327.81
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing