
Hycon® Agar Strips TC-y

Synonym(s):
Hycon Ready-to-use Agar Strips
Products
Product Category
Brand
Physical Form
Agency
Chemical Composition
Compatibility
Quality Segment
Application
Microorganism Suitability
Package Size
Sterilization Method
Showing 1-1 of 1