
Formetanate hydrochloride

Empirical Formula (Hill Notation):
C11H15N3O2 · HCl
CAS Number:
Molecular Weight:
257.72
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Quality Segment
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing