![Fenvalerate certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/860/038/4aff8f1c-cd0c-4610-b90c-2ef008df3a84/640/4aff8f1c-cd0c-4610-b90c-2ef008df3a84.png)
Fenvalerate
![Fenvalerate certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/860/038/4aff8f1c-cd0c-4610-b90c-2ef008df3a84/640/4aff8f1c-cd0c-4610-b90c-2ef008df3a84.png)
Synonym(s):
α-Cyano-3-phenoxybenzyl α-(4-chlorophenyl)isovalerate
Empirical Formula (Hill Notation):
C25H22ClNO3
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Purity
Format
Research Area
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3