
Eppendorf® ep Dualfilter T.I.P.S.®

Synonym(s):
Eppendorf® ep Dualfilter T.I.P.S.®
Products
Product Category
Color
Manufacturer
Binding Type
Volume (ml)
Volume (µL)
Color
Package Size
Sterility
Showing 1-15 of 15
Compare
Product Number
Description
Pricing