
Dulbecco’s Modified Eagle’s Medium/Nutrient Mixture F-12 Ham

Synonym(s):
DME/F-12, 1:1 mixture
Products
Product Category
Brand
Physical Form
HEPES (mM)
L-Glutamine
Sodium Bicarbonate
Phenol Red
Quality Segment
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing