
(±)-Amphetamine-d11 solution

Synonym(s):
(±)-Amphetamine-d11 solution
Empirical Formula (Hill Notation):
C9D11H2N
CAS Number:
Molecular Weight:
146.27
Products
Brand
Formula Weight
Manufacturer
Physical Form
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Technique