
(+)-Isomenthol

Synonym(s):
(1S,2R,5R)-(+)-Isomenthol, (1S,2R,5R)-2-Isopropyl-5-methylcyclohexanol
Empirical Formula (Hill Notation):
C10H20O
CAS Number:
Molecular Weight:
156.27
Products
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing