
Chlorfenapyr

Synonym(s):
4-Bromo-2-(4-chlorophenyl)-1-(ethoxymethyl)-5-(trifluoromethyl)-1H-pyrrol-3-carbonitrile
Empirical Formula (Hill Notation):
C15H11BrClF3N2O
CAS Number:
Molecular Weight:
407.61
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2