Skip to Content
Merck
Bupivacaine hydrochloride monohydrate analytical standard, for drug analysis

Bupivacaine hydrochloride monohydrate

Bupivacaine hydrochloride monohydrate analytical standard, for drug analysis
Synonym(s):
1-Butyl-N-(2,6-dimethylphenyl)-2-piperidinecarboxamide
Empirical Formula (Hill Notation):
C18H28N2O · HCl · H2O
CAS Number:
Molecular Weight:
342.90
EC Number:

Products

Formula Weight
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing