
Bis(3,5-dimethyl-4-methoxyphenyl)chlorophosphine

Empirical Formula (Hill Notation):
C18H22ClO2P
CAS Number:
Molecular Weight:
336.79
Products
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Reaction Type
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing