
Bacopaside II

Synonym(s):
Pseudojujubogenin 3-O-[α-L-arabinofuranosyl-(1→2)-[β-D-glucopyranosyl-(1→3)]-β-D-glucopyranoside]
Empirical Formula (Hill Notation):
C47H76O18
CAS Number:
Molecular Weight:
929.10
Products
Brand
Formula Weight
Manufacturer
Product Line
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing