
Anti-CDHR5 antibody produced in rabbit

Synonym(s):
Anti-μ-Protocadherin, Anti-MUCDHL, Anti-MUPCDH, Anti-Mucin and cadherin-like protein precursor
Products
Product Category
Brand
Biological Source
Clonality
Feature
Gene Alias
Physical Form
Product Line
Antibody Form
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing