
Amantadine Related Compound B

Synonym(s):
N-(1-Adamantyl)acetamide
Empirical Formula (Hill Notation):
C12H19NO
CAS Number:
Molecular Weight:
193.29
Products
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2