Skip to Content
Merck
Allyl methyl carbonate 98%

Allyl methyl carbonate

Allyl methyl carbonate 98%
Linear Formula:
CH2=CHCH2OCOOCH3
CAS Number:
Molecular Weight:
116.12

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Purity
Markush Class
Markush Group
Monomer Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing