Skip to Content
Merck
α-D-Glucose 1,6-bisphosphate potassium salt hydrate ≥99.0% (TLC)

α-D-Glucose 1,6-bisphosphate potassium salt hydrate

α-D-Glucose 1,6-bisphosphate potassium salt hydrate ≥99.0% (TLC)
Synonym(s):
α-D-Glucose 1,6-diphosphate potassium salt hydrate
Empirical Formula (Hill Notation):
C6H10K4O12P2 · xH2O
CAS Number:
Molecular Weight:
492.48 (anhydrous basis)

Products

Product Category
Brand
Biological Source
Color
Formula Weight
Physical Form
Purity
Quality Segment
Optical Isomer