Skip to Content
Merck
4-Hydroxybenzyl alcohol ≥98%, FG

4-Hydroxybenzyl alcohol

4-Hydroxybenzyl alcohol ≥98%, FG
Synonym(s):
NSC 227926
Linear Formula:
HOC6H4CH2OH
CAS Number:
Molecular Weight:
124.14
EC Number:

Products

Product Category
Brand
Biological Source
Formula Weight
Melting Point (°C)
Physical Form
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Reagent Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing