![4arm-PEG5K-Succinimidyl Carboxymethyl Ester average Mn 5,000](/deepweb/assets/sigmaaldrich/product/structures/149/998/a56c6952-83ae-4856-9b00-7382f5332754/640/a56c6952-83ae-4856-9b00-7382f5332754.png)
4arm-PEG5K
![4arm-PEG5K-Succinimidyl Carboxymethyl Ester average Mn 5,000](/deepweb/assets/sigmaaldrich/product/structures/149/998/a56c6952-83ae-4856-9b00-7382f5332754/640/a56c6952-83ae-4856-9b00-7382f5332754.png)
Synonym(s):
4arm-PEG-SH, 4arm-PEG-NH2, 4arm-PEG-Succinimidyl Carboxymethyl Ester
Linear Formula:
C(CH2O(CH2CH2O)nCH2COOC4H4O2N)4
Products
Brand
Physical Form
Functionality
Shape
Polymer Composition
Polymer Type
Reactivity
Showing 1-3 of 3