Key Documents
TMO312751-9050
Nalgene® Dropper Bottle without tip or cap
white low-density polyethylene, volume 15 mL, case of 2000 ea
Synonym(s):
LDPE dropper bottle, dispensing bottle
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
material
white low-density polyethylene
packaging
case of 2000 ea
manufacturer/tradename
Nalgene 312751-9050
capacity
15 mL
volume
15 mL
General description
Repeatedly carry out reliable routine dispensation with Thermo Scientific Nalgene LDPE Dropper Bottles, made of low-density polyethylene. These bottles are to be used with Nalgene controlled dropper tips and closures.
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries