Skip to Content
Merck
All Photos(1)

Key Documents

M9823

Sigma-Aldrich

Myra A hydrochloride

≥98% (HPLC), powder

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H20N2O4 · xHCl
CAS Number:
Molecular Weight:
340.37 (free base basis)
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

storage condition

protect from light

color

red

solubility

DMSO: >10 mg/mL

storage temp.

2-8°C

SMILES string

Cl.CN(C)CCCNc1cc(O)c2C(=O)c3ccccc3C(=O)c2c1O

InChI

1S/C19H20N2O4.ClH/c1-21(2)9-5-8-20-13-10-14(22)15-16(19(13)25)18(24)12-7-4-3-6-11(12)17(15)23;/h3-4,6-7,10,20,22,25H,5,8-9H2,1-2H3;1H

InChI key

ZVKHFHKBUHMRPU-UHFFFAOYSA-N

Biochem/physiol Actions

Myra A hydrochloride is a small molecule transcription factor inhibitor (c-Myc/Max DNA binding).

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service