Skip to Content
Merck
All Photos(1)

Key Documents

XX1104707

Millipore

O-ring, (5-329) for Swinnex 47 mm filter holder, silicone

Accessories for filter holders for sample preparation.

Synonym(s):

Replacement O-ring, Silicone O-ring

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104909
eCl@ss:
32031613
NACRES:
NB.24

material

silicone O-ring

Quality Level

diam.

47 mm

compatibility

for use with Swinnex 47

shipped in

ambient

General description

Accessories for filter holders for sample preparation.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service