Key Documents
NA38/150
Millipore
NovAseptic® Valve, Manual Plastic Actuator
About This Item
UNSPSC Code:
40141600
eCl@ss:
37051001
material
plastic handle (Oilon)
stainless steel cover (EN 1.4305/ASTM 303)
stainless steel indicator (SS 2346)
Quality Level
product line
NovAseptic®
parameter
pressure range at -1-10 bar
temp. range at 0-70 °C
weight
1.2 kg (2.65 lb)
Application
The applied valve body and diaphragm may have different design temperature and/or pressure limits. The weakest component in the assembled product determines the maximum design temperature and pressure limits.
Other Notes
Actuator, Manual
Legal Information
NOVASEPTIC is a registered trademark of Merck KGaA, Darmstadt, Germany