Skip to Content
Merck
All Photos(1)

Key Documents

1.16216

Sigma-Aldrich

Peroxidase

(from horse radish) 10000 U/vial for biochemistry EC 1.11.1.7

Synonym(s):

Peroxidase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352204
EC Index Number:
232-668-6
NACRES:
NA.28

biological source

horseradish

Quality Level

form

solid

pH

6.5-7.5 (20 °C, 10 g/L in H2O)

storage temp.

2-8°C

InChI

1S/H2O3/c1-3-2/h1-2H

InChI key

JSPLKZUTYZBBKA-UHFFFAOYSA-N

General description

Horseradish peroxidase (HRP) is a glycoprotein that belongs to the class of oxidoreductase of the peroxidases family. It is found in horseradish roots. HRP contains significant amounts of carbohydrates and photohemin IX as its prosthetic group.

Biochem/physiol Actions

Peroxidases catalyze the cleavage of peroxides by redox reactions. The active center includes heme, cysteine thiols, selenium, manganese, and other chemical moieties. Peroxidase isoenzymes are essential for oxidizing reactive oxygen species, innate immunity, hormone biosynthesis, and the pathogenesis of many diseases. They are generally used for sensors, drug delivery devices, environment protection, bioremediation, and energy production. Horseradish peroxidase (HRP) is the widely used oxidoreductase in colorimetric, fluorimetric, chemiluminescent, electrochemical, and immunoassays. Its concentration can be measured with O-phenylenediamine (OPD) and hydrogen peroxide. HRP serves as a tracer for immunocytochemistry and is widely used in antibody-enzyme conjugates.

Features and Benefits

  • Ideal for preparing small-sized antibody-enzyme conjugates
  • Enables better cellular penetration through low-molecular-weight conjugates
  • Small conjugate size ensures greater accessibility to antigenic structures within tissue sections
  • Stable even under the conditions employed for crosslinking

Analysis Note

Activity
- Hydrogen peroxide and guajacol; pH 7.0; 25 °C:
about 170 U/mg
: ≥ 10000 U/Glas
Catalase: ≤ 1 U/mg

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service