Key Documents
Z556750
Sigma-Aldrich
Aldrich® swivel Benchclamp
2-prong, size M
Synonym(s):
bench clamps
About This Item
UNSPSC Code:
41122413
NACRES:
NB.35
size
M
jaw capacity
0-76 mm
prong W
23 mm
General description
Aldrich benchclamp clamps have a nickel-plated finish and are corrosion resistant. The clamps come with slip-on fiberglass finger covers. The Grippy Knob design has a round nut which is easy to turn making tightening the clamps around glassware fast and convenient. This swivel head allows for the clamp to have any angle of adjustment.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Benchclamp is a trademark of Sigma-Aldrich Co. LLC