MT51350004
Mettler-Toledo technical buffer
pH 4.01, volume 250 mL
Synonym(s):
liquid pH 4.01 buffer
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
51191600
NACRES:
NB.79
Recommended Products
sterility
non-sterile
form
liquid
manufacturer/tradename
Mettler-Toledo 51350004
volume
250 mL
pH
4.01
Looking for similar products? Visit Product Comparison Guide
General description
METTLER TOLEDO certified pH buffer solutions are traceable to NIST primary standards and come with a quality inspection certificate, which guarantees the stated values and traceability. We offer convenient package sizes from single-use 20 mL pouches to 20 L Cubitainers. Our variety packs of pH 4, 7 and 10 buffers are offered in both 20 mL pouches and convenient 500 mL bottles.
Features and Benefits
- High-quality Mettler-Toledo Rainbow I buffers for accurate sensor calibration and maintenance.
- Small volume packaging (single-use sachets) ensures guaranteed fresh solutions and ease of use.
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service