Skip to Content
Merck
All Photos(1)

Key Documents

M30507

Sigma-Aldrich

Methyl benzoylformate

98%

Synonym(s):

Methyl phenylglyoxylate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5COCOOCH3
CAS Number:
Molecular Weight:
164.16
Beilstein:
1100868
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

98%

form

liquid

refractive index

n20/D 1.526 (lit.)

bp

246-248 °C (lit.)

density

1.155 g/mL at 25 °C (lit.)

SMILES string

COC(=O)C(=O)c1ccccc1

InChI

1S/C9H8O3/c1-12-9(11)8(10)7-5-3-2-4-6-7/h2-6H,1H3

InChI key

YLHXLHGIAMFFBU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Roman Tsaryk et al.
Journal of tissue engineering and regenerative medicine, 11(3), 637-648 (2014-11-06)
Limitations of current treatments for intervertebral disc (IVD) degeneration have promoted interest in the development of tissue-engineering approaches. Injectable hydrogels loaded with cells can be used as a substitute material for the inner IVD part, the nucleus pulposus (NP), and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service