HPLC Analysis of Positional Isomers (Xylenes) on Astec® CYCLOBOND I 2000
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
CYCLOBOND I 2000, 25 cm x 4.6 mm I.D., 5 μm particles (20024AST)
mobile phase
[A] acetonitrile; [B] water; (15:85, A:B)
flow rate
0.8 mL/min
column temp.
55 °C
detector
UV, 230 nm
injection
3 μL
sample
xylene isomers, each compound, 0.1 mg/mL in acetonitrile:water (50:50) (elution order: m, o, p)
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
null