HPLC Analysis of MAb 016 on Proteomix® SCX NP5 versus Competitive SCX using a Salt Gradient
Materials
analytical column
Product No.
Description
Pricing
Proteomix® SCX-NP5 Strong Cation Exchange Column
L × I.D. 25 cm × 4.6 mm, 5 μm particle size, PEEKCONDITIONS
column
Proteomix SCX-NP5, 25 cm x 4.6 mm I.D. (PEEK hardware), 5 μm particles (Z777164)
mobile phase
(A) 20 mM phosphate buffer, pH 7.5; (B) A + 1 M NaCl
gradient
0 to 6% B in 30 min
flow rate
0.8 mL/min
detector
UV, 280 nm
injection
4 μL
sample
MAb 016 (11.8 mg/mL)
Description
Legal Information
Proteomix is a registered trademark of Sepax Technologies