Merck
  • Oxidative DNA damage contributes to the toxic activity of propylparaben in mammalian cells.
Oxidative DNA damage contributes to the toxic activity of propylparaben in mammalian cells.
Mutation research (2010-08-05)
José Manuel Pérez Martín, Ana Peropadre, Oscar Herrero, Paloma Fernández Freire, Verónica Labrador, María José Hazen
ABSTRACT
Propyl p-hydroxybenzoate, commonly referred to as propylparaben, is the most frequently used preservative to inhibit microbial growth and extend shelf life of a range of consumer products. The objective of this study was to provide further insight into the toxicological profile of this compound, because of the current discrepancy in the literature with regard to the safety of parabens. The Vero cell line, derived from the kidney of the green monkey, was selected to evaluate the adverse effects of propylparaben by use of a set of mechanistically relevant endpoints for detecting cytotoxicity and genotoxic activities. Our results demonstrate that exposure to the compound for 24h causes changes in cell-proliferation rates rather than in cell viability. A significant and dose-dependent decline in the percentage of mitotic cells was observed at the lowest concentration tested, mainly due to cell-cycle arrest at the G0/G1 phase. Immunodetection techniques revealed that induction of DNA double-strand breaks and oxidative damage underlies the cytostatic effect observed in treated Vero cells. Additional studies are in progress to extend these findings, which define a novel mode of action of propylparaben in cultured mammalian cells.
Product Description
Propyl parahydroxybenzoate, European Pharmacopoeia (EP) Reference Standard
Supelco
Propylparaben, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Propyl 4-hydroxybenzoate, ≥99%
Sigma-Aldrich
Propyl 4-hydroxybenzoate, tested according to Ph. Eur.
Sigma-Aldrich
Propyl 4-hydroxybenzoate, BioXtra