Merck
  • Antitumor activity of Ru(III) complexes carrying beta-diketonato ligands in vitro and in vivo.
Antitumor activity of Ru(III) complexes carrying beta-diketonato ligands in vitro and in vivo.
Journal of B.U.ON. : official journal of the Balkan Union of Oncology (2009-08-04)
S S Arandjelovic, K S Bjelogrlic, N N Malesevic, Lj Z Tesic, S S Radulovic
ABSTRACT
To investigate the antitumor activity of two newly synthesized ruthenium(III) [Ru(III)] compounds carrying bidentate ligands: (acac)-acetylacetonate, [Ru(acac)3), and (tfac)-trifluoroacetylacetonate [Ru(tfac)3]. The activity of ruthenium(III) analogues was evaluated on HeLa, B16, and Femx cell lines for cytotoxicity in vitro using MTT assay, and inhibition on tumor invading ability in vitro using cell migration and invasion assays, whereas inhibition of tumor growth in vivo was estimated on advanced B16 murine melanoma model. Both compounds were also investigated in combinations with cisplatin, oxaliplatin, or poly ADP-ribose polymerase- 1 (PARP-1) inhibitor, in order to determine the pattern of mutual interactions. Applied as single drugs, Ru(tfac)3 showed high cytotoxic activity against HeLa and Femx cell lines, while Ru(acac)3 did not reach the IC50 on any of the cell lines tested. In combinations, Ru(acac)3 with cisplatin gained synergistic interaction, antagonistic with oxaliplatin, and of different kind with (PARP-1) inhibitor in concentration-and cell line-dependent manner. Ru(acac)3 exhibited inhibition of HeLa cell migration and gelatinolytic activity of MMP-2 and MMP-9. Ru(tfac)3 complexes did not induce significant reduction of melanoma growth in vivo, whereas Ru(acac)3 did, but the latter failed to contribute in lifespan improvement. The investigated ruthenium complexes showed different levels of antitumor activity in vitro and in vivo, implicating on different mechanisms of their action as well as diverse perspectives in cancer treatment.
Sigma-Aldrich
Copper(II) acetylacetonate, ≥99.9% trace metals basis
Sigma-Aldrich
Copper(II) acetylacetonate, 97%
Sigma-Aldrich
Manganese(III) acetylacetonate, technical grade
Sigma-Aldrich
Manganese(II) acetylacetonate
Sigma-Aldrich
Chromium(III) acetylacetonate, 97%