Merck
  • Quantitation of bergenin in human plasma by liquid chromatography/tandem mass spectrometry.
Quantitation of bergenin in human plasma by liquid chromatography/tandem mass spectrometry.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2008-11-28)
Weisong Yu, Yingwu Wang, Yunhui Zhang, Dan Zhang, Jing Lan, Zeyuan Liu, Jingkai Gu, J Paul Fawcett
ABSTRACT
This paper reports the development and validation of an assay for quantitation of bergenin in human plasma using liquid chromatography/tandem mass spectrometry (LC-MS/MS). Bergenin and the internal standard (I.S.), 5-bromo-2,4(1H,3H)-pyrimidinedione (5-BrU), were separated by reversed phase HPLC and quantitated by MS/MS using electrospray ionization (ESI) and multiple reaction monitoring (MRM) in the negative ion mode. The most intense [M-H](-) MRM transition of bergenin at m/z 326.9-->312.3 was used for quantitation and the transition at m/z 188.9-->42.2 was used to monitor 5-BrU. Stability issues with bergenin required the addition of ascorbic acid to plasma samples prior to storage and analysis within 10 days storage at -80 degrees C. The method was linear in the range 3-1000 ng/mL with intra- and inter-day precision of 3.94-5.96 and 1.62-8.31%, respectively, and accuracy <2.33%. The assay was successfully applied to a pharmacokinetic study in healthy volunteers after administration of a single 250 mg oral dose.
Sigma-Aldrich
Bergenin, ≥95% (LC/MS-ELSD)