Merck
L-amino acid oxidase-induced apoptosis in filamentous Botrytis cinerea.
Analytical biochemistry (2011-09-29)
Chi-Hua Cheng, Chia-Ann Yang, Shu-Ying Liu, Chaur-Tsuen Lo, Kou-Cheng Peng
ABSTRACT
As opposed to single-cell yeast and mammalian cell lines, apoptosis has not been greatly investigated in filamentous fungi because antibodies to the relevant fungal apoptosis-related proteins are not available commercially and because multicellular organisms cannot be studied using flow cytometry. Here we demonstrate how antibodies from a nonfungal source could be used to investigate this pathway. We show that apoptosis in the filamentous fungus Botrytis cinerea is triggered by the mitochondria-mediated caspase pathway, with release of the apoptotic factors cytochrome c, caspase 3, and caspase 9, on treatment with Trichoderma harzianum-derived L-amino acid oxidase.
Sigma-Aldrich
L-Amino Acid Oxidase from Crotalus adamanteus, Type I (dried venom)
Sigma-Aldrich
L-Amino Acid Oxidase from Crotalus atrox (Western Diamondback Rattlesnake), Type VI, dried venom
Sigma-Aldrich
L-Amino Acid Oxidase from Crotalus adamanteus, Type IV, ≥4.0 units/mg protein, aqueous suspension