Merck
  • Synergistic cooperation between hypoxia and transforming growth factor-beta pathways on human vascular endothelial growth factor gene expression.
Synergistic cooperation between hypoxia and transforming growth factor-beta pathways on human vascular endothelial growth factor gene expression.
The Journal of biological chemistry (2001-08-04)
T Sánchez-Elsner, L M Botella, B Velasco, A Corbí, L Attisano, C Bernabéu
ABSTRACT
Signaling by transforming growth factor (TGF)-beta family members is mediated by Smad proteins that regulate gene transcription through functional cooperativity and association with other DNA-binding proteins. The hypoxia-inducible factor (HIF)-1 is a transcriptional complex that plays a key role in oxygen-regulated gene expression. We demonstrate that hypoxia and TGF-beta cooperate in the induction of the promoter activity of vascular endothelial growth factor (VEGF), which is a major stimulus in the promotion of angiogenesis. This cooperation has been mapped on the human VEGF promoter within a region at -1006 to -954 that contains functional DNA-binding sequences for HIF-1 and Smads. Optimal HIF-1alpha-dependent induction of the VEGF promoter was obtained in the presence of Smad3, suggesting an interaction between these proteins. Consistent with this, co-immunoprecipitation experiments revealed that HIF-1alpha physically associates with Smad3. These results demonstrate that both TGF-beta and hypoxia signaling pathways can synergize in the regulation of VEGF gene expression at the transcriptional level.
Sigma-Aldrich
Deferoxamine mesylate salt, powder, ≥92.5% (TLC)
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)