Merck
  • Sensory and Volatile Profiles of Korean Commercially Distilled Soju Using Descriptive Analysis and HS-SPME-GC-MS.
Sensory and Volatile Profiles of Korean Commercially Distilled Soju Using Descriptive Analysis and HS-SPME-GC-MS.
Foods (Basel, Switzerland) (2020-09-25)
Jung-Min Hong, Tae-Wan Kim, Seung-Joo Lee
ABSTRACT
Volatile compositions and sensory characteristics of 11 commercially distilled soju samples were investigated using headspace solid-phase microextraction (HS-SPME) with gas chromatography-mass spectrometry (GC-MS) and sensory descriptive analysis. A total of 59 major volatile compounds, consisting of 32 esters, 10 alcohols, 2 acids, 5 aldehydes, 3 ketones, 1 hydrocarbon, 1 furan, 2 phenols, and 3 miscellaneous compounds, were identified. From the principal component analysis (PCA) of volatile data, MSJ made by atmospheric distillation showed a clear distinction in volatile compositions compared to that of other samples made by vacuum distillation. Based on PCA of the sensory data determined by a panel of ten judges, MSJ was associated with a large amount of longer chain esters that showed high intensities in bitter taste and yeast/nuruk-related flavor attributes. HYJ, LPJ, and HAJ made with rice as a raw material were associated with lower intensities of the alcohol aroma, while JRJ and OKJ aged in oak barrels were associated with fruit flavor, sweet flavor, and brandy aroma. In the partial least squares regression (PLSR) analysis to see any relationship between volatile and sensory data, longer chain esters like ethyl tetradecanoate, and ethyl hexadecanoate were highly associated with bleach aroma. In contrast, positive correlations were seen with barley aroma and yeast flavor with hexanal, nonanal, benzaldehyde, and 2-methoxy-phenol.
Sigma-Aldrich
2-Methyl-1-pentanol, 99%
Sigma-Aldrich
Suplatast tosylate, ≥98% (HPLC)