Merck
  • Expanded Satellite Repeats Amplify a Discrete CENP-A Nucleosome Assembly Site on Chromosomes that Drive in Female Meiosis.
Expanded Satellite Repeats Amplify a Discrete CENP-A Nucleosome Assembly Site on Chromosomes that Drive in Female Meiosis.
Current biology : CB (2017-08-02)
Aiko Iwata-Otsubo, Jennine M Dawicki-McKenna, Takashi Akera, Samantha J Falk, Lukáš Chmátal, Karren Yang, Beth A Sullivan, Richard M Schultz, Michael A Lampson, Ben E Black
ABSTRACT
Female meiosis provides an opportunity for selfish genetic elements to violate Mendel's law of segregation by increasing the chance of segregating to the egg [1]. Centromeres and other repetitive sequences can drive in meiosis by cheating the segregation process [2], but the underlying mechanisms are unknown. Here, we show that centromeres with more satellite repeats house more nucleosomes that confer centromere identity, containing the histone H3 variant CENP-A, and bias their segregation to the egg relative to centromeres with fewer repeats. CENP-A nucleosomes predominantly occupy a single site within the repeating unit that becomes limiting for centromere assembly on smaller centromeres. We propose that amplified repetitive sequences act as selfish elements by promoting expansion of CENP-A chromatin and increased transmission through the female germline.
Sigma-Aldrich
Nocodazole, ≥99% (TLC), powder
Roche
Anti-Digoxigenin-Fluorescein, Fab fragments, from sheep
Roche
Nick Translation Kit, sufficient for 50 labeling reactions, kit of 1 (7 components), suitable for FISH