Merck
  • Combined Targeting of Estrogen Receptor Alpha and Exportin 1 in Metastatic Breast Cancers.
Combined Targeting of Estrogen Receptor Alpha and Exportin 1 in Metastatic Breast Cancers.
Cancers (2020-08-28)
Eylem Kulkoyluoglu Cotul, Qianying Zuo, Ashlie Santaliz-Casiano, Ozan Berk Imir, Ayca Nazli Mogol, Elif Tunc, Kevin Duong, Jenna Kathryn Lee, Rithva Ramesh, Elijah Odukoya, Mrinali P Kesavadas, Monika Ziogaite, Brandi Patrice Smith, Chengjian Mao, David J Shapiro, Ben Ho Park, Benita S Katzenellenbogen, Drew Daly, Evelyn Aranda, John D O'Neill, Christopher Walker, Yosef Landesman, Zeynep Madak-Erdogan
ABSTRACT
The majority of breast cancer specific deaths in women with estrogen receptor positive (ER+) tumors occur due to metastases that are resistant to therapy. There is a critical need for novel therapeutic approaches to achieve tumor regression and/or maintain therapy responsiveness in metastatic ER+ tumors. The objective of this study was to elucidate the role of metabolic pathways that undermine therapy efficacy in ER+ breast cancers. Our previous studies identified Exportin 1 (XPO1), a nuclear export protein, as an important player in endocrine resistance progression and showed that combining selinexor (SEL), an FDA-approved XPO1 antagonist, synergized with endocrine agents and provided sustained tumor regression. In the current study, using a combination of transcriptomics, metabolomics and metabolic flux experiments, we identified certain mitochondrial pathways to be upregulated during endocrine resistance. When endocrine resistant cells were treated with single agents in media conditions that mimic a nutrient deprived tumor microenvironment, their glutamine dependence for continuation of mitochondrial respiration increased. The effect of glutamine was dependent on conversion of the glutamine to glutamate, and generation of NAD+. PGC1α, a key regulator of metabolism, was the main driver of the rewired metabolic phenotype. Remodeling metabolic pathways to regenerate new vulnerabilities in endocrine resistant breast tumors is novel, and our findings reveal a critical role that ERα-XPO1 crosstalk plays in reducing cancer recurrences. Combining SEL with current therapies used in clinical management of ER+ metastatic breast cancer shows promise for treating and keeping these cancers responsive to therapies in already metastasized patients.
Sigma-Aldrich
Anti-PGC1A antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
L-(−)-Glucose, ≥99%
Sigma-Aldrich
MONOCLONAL ANTI-BETA-ACTIN antibody produced in mouse, clone 8H10D10, IgG fraction of antiserum, buffered aqueous solution