Merck
HIGHER-ORDER ABERRATIONS IN EYES WITH SILICONE OIL TAMPONADE.
Retina (Philadelphia, Pa.) (2019-01-15)
Joo Youn Shin, In Kwon Chung, Ga Hee Han, Suk Ho Byeon, Jin Hyoung Kim, Doh Lee, Jonghyun Lee
ABSTRACT
To investigate the changes in higher-order aberrations (HOAs) after silicone oil removal and to evaluate their associations with visual acuity. Fifty-nine eyes of 58 patients who underwent SO removal were included. Total, corneal, and internal optic HOAs, and best-corrected visual acuity were measured before and 1 month after SO removal, and changes were compared between phakic and pseudophakic eyes. Total ocular and internal optic HOAs decreased significantly after SO removal both in pseudophakic (n = 40, all P < 0.001) and phakic eyes (n = 19, P = 0.017, P = 0.004). Preoperative HOAs (P < 0.001) and changes in HOAs (P = 0.006) were greater in pseudophakic eyes than in phakic eyes. Best-corrected visual acuity was significantly improved after SO removal, from 20/105 to 20/78 (P < 0.001) in pseudophakic eyes, whereas there was no difference in phakic eyes (P = 0.714). Preoperative HOAs and the reduction in HOAs after SO removal were greater in best-corrected visual acuity-improved eyes than best-corrected visual acuity-unchanged eyes (P < 0.001). Silicone oil tamponade induced an increase in HOAs, and these increases were greater in pseudophakic eyes than in phakic eyes. Silicone oil may cause additional visual impairments because of HOAs, beyond those caused by retinal diseases, particularly in pseudophakic eyes.
Sigma-Aldrich
Tetrahydropyran, anhydrous, 99%