Merck
  • CD97, an adhesion receptor on inflammatory cells, stimulates angiogenesis through binding integrin counterreceptors on endothelial cells.
CD97, an adhesion receptor on inflammatory cells, stimulates angiogenesis through binding integrin counterreceptors on endothelial cells.
Blood (2004-12-04)
Tao Wang, Yvona Ward, Linhua Tian, Ross Lake, Liliana Guedez, William G Stetler-Stevenson, Kathleen Kelly
ABSTRACT
CD97, a membrane protein expressed at high levels on inflammatory cells and some carcinomas, is a member of the adhesion G protein-coupled receptor family, whose members have bipartite structures consisting of an extracellular peptide containing adhesion motifs noncovalently coupled to a class B 7-transmembrane domain. CD97alpha, the extracellular domain of CD97, contains 3 to 5 fibrillin class 1 epidermal growth factor (EGF)-like repeats, an Arg-Gly-Asp (RGD) tripeptide, and a mucin stalk. We show here that CD97alpha promotes angiogenesis in vivo as demonstrated with purified protein in a directed in vivo angiogenesis assay (DIVAA) and by enhanced vascularization of developing tumors expressing CD97. These data suggest that CD97 can contribute to angiogenesis associated with inflammation and tumor progression. Strong integrin alpha5beta1 interactions with CD97 have been identified, but alpha v beta3 also contributes to cell attachment. Furthermore, soluble CD97 acts as a potent chemoattractant for migration and invasion of human umbilical vein endothelial cells (HUVECs), and this function is integrin dependent. CD97 EGF-like repeat 4 is known to bind chondroitin sulfate. It was found that coengagement of alpha5beta1 and chondroitotin sulfate proteoglycan by CD97 synergistically initiates endothelial cell invasion. Integrin alpha5beta1 is the first high-affinity cellular counterreceptor that has been identified for a member within this family of adhesion receptors.
Sigma-Aldrich
Anti-Integrin αVβ3 Antibody, clone LM609, clone LM609, Chemicon®, from mouse
Sigma-Aldrich
Anti-Integrin αVβ5 Antibody, clone P1F6, clone P1F6, Chemicon®, from mouse
Sigma-Aldrich
Anti-Integrin α2 Antibody, clone P1E6, clone P1E6, Chemicon®, from mouse